Assay Details
Gene Symbol: PRSS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266588.1 | NP_001253517.1 | 5 - 6 | 722 | 147 | |
XM_015125992.2 | XP_014981478.1 | 1162 | 147 | |||
GenBank mRNA | JU476439.1 | - | 5 - 6 | 722 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
