Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194028.1 | NP_001180957.1 | 7 - 8 | 819 | 74 | |
GenBank mRNA | JU476476.1 | - | 7 - 8 | 814 | 74 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
