Assay Details
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117605.4 | XP_001117605.1 | 777 | 80 | ||
GenBank mRNA | JU470789.1 | - | 756 | 80 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
