Assay Details
Gene Symbol: ZNF282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098866.4 | XP_001098866.3 | 288 | 58 | ||
GenBank mRNA | JU324888.1 | - | 1 - 2 | 233 | 58 | |
JV663337.1 | - | 1 - 2 | 251 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
