Assay Details
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105622.4 | XP_001105622.4 | 284 | 66 | ||
XM_028832373.1 | XP_028688206.1 | 284 | 66 | |||
GenBank mRNA | JU349546.1 | - | 151 | 66 | ||
JU477510.1 | - | 266 | 66 | |||
JV048458.1 | - | 259 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
