Assay Details
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260579.1 | NP_001247508.1 | 1956 | 66 |
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257871.1 | NP_001244800.1 | 1060 | 66 | ||
XM_015130262.2 | XP_014985748.2 | 1125 | 66 | |||
XM_015130263.2 | XP_014985749.2 | 997 | 66 | |||
GenBank mRNA | JU323226.1 | - | 1956 | 66 | ||
JU336824.1 | - | 1060 | 66 | |||
JV048517.1 | - | 938 | 66 | |||
JV699453.1 | - | 1094 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
