Assay Details
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260579.1 | NP_001247508.1 | 2779 | 126 |
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257871.1 | NP_001244800.1 | 237 | 126 | ||
XM_015130262.2 | XP_014985748.2 | 302 | 126 | |||
GenBank mRNA | JU323226.1 | - | 2779 | 126 | ||
JU336824.1 | - | 237 | 126 | |||
JV048517.1 | - | 115 | 126 | |||
JV699453.1 | - | 271 | 126 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
