Assay Details
Gene Symbol: OGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260579.1 | NP_001247508.1 | 2435 | 72 |
Gene Symbol: CAMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257871.1 | NP_001244800.1 | 581 | 72 | ||
XM_015130262.2 | XP_014985748.2 | 646 | 72 | |||
XM_015130263.2 | XP_014985749.2 | 518 | 72 | |||
GenBank mRNA | JU323226.1 | - | 2435 | 72 | ||
JU336824.1 | - | 581 | 72 | |||
JV048517.1 | - | 459 | 72 | |||
JV699453.1 | - | 615 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
