Assay Details
Gene Symbol: DEGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266006.1 | NP_001252935.1 | 1 - 2 | 152 | 102 | |
GenBank mRNA | JU322768.1 | - | 1 - 2 | 186 | 102 | |
JU332241.1 | - | 1 - 2 | 186 | 102 | ||
JU332242.1 | - | 1 - 2 | 186 | 102 | ||
JU332243.1 | - | 1 - 2 | 186 | 102 | ||
JU332244.1 | - | 1 - 2 | 186 | 102 | ||
JU332245.1 | - | 1 - 2 | 186 | 102 | ||
JU332246.1 | - | 1 - 2 | 186 | 102 | ||
JU332247.1 | - | 1 - 2 | 186 | 102 | ||
JU332248.1 | - | 1 - 2 | 186 | 102 | ||
JU332249.1 | - | 1 - 2 | 186 | 102 | ||
JU471089.1 | - | 1 - 2 | 160 | 102 | ||
JV045802.1 | - | 1 - 2 | 152 | 102 | ||
JV699644.1 | - | 1 - 2 | 188 | 102 | ||
JV725811.1 | - | 1 - 2 | 186 | 102 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
