Assay Details
Gene Symbol: CASK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127187.2 | XP_014982673.1 | 1911 | 128 | ||
XM_015127188.2 | XP_014982674.1 | 1912 | 128 | |||
XM_015127189.2 | XP_014982675.1 | 1893 | 128 | |||
XM_015127190.2 | XP_014982676.1 | 1893 | 128 | |||
XM_015127191.2 | XP_014982677.1 | 1759 | 128 | |||
XM_015127192.2 | XP_014982678.1 | 1874 | 128 |
Gene Symbol: LOC114675624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842839.1 | XP_028698672.1 | 583 | 128 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
