Assay Details
Gene Symbol: NPFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106733.3 | XP_001106733.1 | 839 | 123 | ||
XM_015151876.2 | XP_015007362.1 | 1069 | 123 | |||
XM_028829571.1 | XP_028685404.1 | 998 | 123 | |||
XM_028829572.1 | XP_028685405.1 | 1200 | 123 | |||
GenBank mRNA | JU325182.1 | - | 93 | 123 | ||
JV074276.1 | - | 44 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
