Assay Details
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261693.1 | NP_001248622.1 | 5 - 6 | 480 | 113 | |
XM_015122994.1 | XP_014978480.1 | 671 | 113 | |||
GenBank mRNA | JU336890.1 | - | 5 - 6 | 475 | 113 | |
JU477631.1 | - | 5 - 6 | 480 | 113 | ||
JV664368.1 | - | 3 - 4 | 236 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
