Assay Details
Gene Symbol: STX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261693.1 | NP_001248622.1 | 6 - 7 | 590 | 98 | |
GenBank mRNA | JU336890.1 | - | 6 - 7 | 585 | 98 | |
JU477631.1 | - | 6 - 7 | 590 | 98 | ||
JV664368.1 | - | 4 - 5 | 346 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
