Assay Details
Gene Symbol: FGF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194186.1 | NP_001181115.1 | 3 - 4 | 399 | 158 | |
XM_001101524.3 | XP_001101524.1 | 752 | 158 | |||
XM_015144891.2 | XP_015000377.1 | 757 | 158 | |||
XM_028852245.1 | XP_028708078.1 | 1113 | 158 |

More Information
Set Membership: |
![]() ![]() ![]() |
