Assay Details
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151162.2 | XP_015006648.1 | 883 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015151162.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
