Assay Details
Gene Symbol: LOC717142 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015116665.2 | XP_014972151.2 | 80 | |||
XM_015116666.2 | XP_014972152.1 | 80 | ||||
XM_015116667.2 | XP_014972153.1 | 80 | ||||
XM_015116668.2 | XP_014972154.1 | 80 | ||||
XM_028842759.1 | XP_028698592.1 | 80 | ||||
GenBank mRNA | JU473411.1 | - | 2 - 3 | 102 | 80 | |
JV045533.1 | - | 2 - 3 | 88 | 80 | ||
JV453234.1 | - | 2 - 3 | 80 | |||
JV527769.1 | - | 2 - 3 | 122 | 80 | ||
JV568008.1 | - | 2 - 3 | 80 |
Gene Symbol: KYAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028834738.1 | XP_028690571.1 | 701 | 80 | ||
XM_028834739.1 | XP_028690572.1 | 404 | 80 | |||
XM_028834740.1 | XP_028690573.1 | 264 | 80 | |||
XM_028834741.1 | XP_028690574.1 | 500 | 80 | |||
XM_028834742.1 | XP_028690575.1 | 149 | 80 | |||
GenBank mRNA | JV601807.1 | - | 2 - 3 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
