Assay Details
Gene Symbol: EEF1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261342.1 | NP_001248271.1 | 3 - 4 | 426 | 140 | |
GenBank mRNA | JU337206.1 | - | 3 - 4 | 423 | 140 | |
JV045565.1 | - | 3 - 4 | 426 | 140 | ||
JV489808.1 | - | 3 - 4 | 452 | 140 | ||
JV598990.1 | - | 3 - 4 | 377 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
