Assay Details
Gene Symbol: MTRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266680.1 | NP_001253609.1 | 423 | 99 | ||
XM_015120894.2 | XP_014976380.1 | 667 | 99 | |||
XM_028837108.1 | XP_028692941.1 | 733 | 99 | |||
GenBank mRNA | JU337240.1 | - | 433 | 99 | ||
JU477952.1 | - | 436 | 99 | |||
JV043258.1 | - | 423 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
