Assay Details
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265676.1 | NP_001252605.1 | 6 - 7 | 1906 | 109 | |
XM_028844719.1 | XP_028700552.1 | 1977 | 109 | |||
GenBank mRNA | JV048457.1 | - | 6 - 7 | 1786 | 109 | |
JV636412.1 | - | 6 - 7 | 1906 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
