Assay Details
Gene Symbol: MAP3K21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265676.1 | NP_001252605.1 | 9 - 10 | 2934 | 150 | |
GenBank mRNA | JV048457.1 | - | 9 - 10 | 2814 | 150 | |
JV636412.1 | - | 9 - 10 | 2934 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
