Assay Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092746.4 | XP_001092746.3 | 306 | 83 | ||
XM_015145198.2 | XP_015000684.1 | 527 | 83 | |||
XM_015145201.2 | XP_015000687.1 | 323 | 83 | |||
GenBank mRNA | JV725215.1 | - | 2 - 3 | 167 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
