Assay Details
Gene Symbol: PGBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099438.4 | XP_001099438.2 | 1770 | 157 | ||
XM_001099547.4 | XP_001099547.2 | 1750 | 157 | |||
XM_015135798.2 | XP_014991284.2 | 3608 | 157 | |||
GenBank mRNA | JU323348.1 | - | 718 | 157 | ||
JV663569.1 | - | 727 | 157 | |||
JV704499.1 | - | 540 | 157 |

More Information
Set Membership: |
![]() ![]() |
