Assay Details
Gene Symbol: FAXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266112.1 | NP_001253041.1 | 2 - 3 | 502 | 78 | |
XM_028846620.1 | XP_028702453.1 | 938 | 78 | |||
GenBank mRNA | JU336750.1 | - | 2 - 3 | 502 | 78 | |
JV631134.1 | - | 2 - 3 | 643 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
