Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266922.1 | NP_001253851.1 | 3 - 4 | 430 | 60 | |
GenBank mRNA | JU336055.1 | - | 3 - 4 | 509 | 60 | |
JV048022.1 | - | 3 - 4 | 430 | 60 | ||
JV659564.1 | - | 3 - 4 | 514 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
