Assay Details
Gene Symbol: GPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138378.2 | XP_014993864.1 | 1084 | 107 | ||
XM_015138380.2 | XP_014993866.1 | 1122 | 107 | |||
XM_028848448.1 | XP_028704281.1 | 1225 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
