Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114142.2 | XP_014969628.2 | 1078 | 113 | ||
XM_015114143.2 | XP_014969629.2 | 756 | 113 | |||
GenBank mRNA | JU327536.1 | - | 395 | 113 | ||
JU477333.1 | - | 708 | 113 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
