Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114142.2 | XP_014969628.2 | 1494 | 149 | ||
XM_015114143.2 | XP_014969629.2 | 1169 | 149 | |||
GenBank mRNA | JU327536.1 | - | 8 - 9 | 811 | 149 | |
JU477333.1 | - | 8 - 9 | 1124 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
