Assay Details
Gene Symbol: ZFP91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114142.2 | XP_014969628.2 | 1704 | 89 | ||
XM_015114143.2 | XP_014969629.2 | 1379 | 89 | |||
GenBank mRNA | JU327536.1 | - | 1021 | 89 | ||
JU477333.1 | - | 1334 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
