Assay Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107704.3 | XP_001107704.1 | 932 | 78 | ||
GenBank mRNA | JU496263.1 | - | 515 | 78 | ||
JV075521.1 | - | 208 | 78 |
