Assay Details
Gene Symbol: MAP3K20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086798.4 | XP_001086798.4 | 774 | 122 | ||
XM_015110384.2 | XP_014965870.2 | 822 | 122 | |||
XM_015110385.2 | XP_014965871.2 | 627 | 122 | |||
GenBank mRNA | JU320852.1 | - | 6 - 7 | 630 | 122 | |
JU475983.1 | - | 7 - 8 | 714 | 122 | ||
JU475984.1 | - | 7 - 8 | 714 | 122 | ||
JU475985.1 | - | 7 - 8 | 714 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
