Assay Details
Gene Symbol: SP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266924.1 | NP_001253853.1 | 4 - 5 | 1888 | 86 | |
XM_015151801.2 | XP_015007287.1 | 1816 | 86 | |||
GenBank mRNA | JU336057.1 | - | 4 - 5 | 1936 | 86 | |
JU476915.1 | - | 4 - 5 | 1926 | 86 | ||
JV048023.1 | - | 4 - 5 | 1888 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
