Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015153020.2 | XP_015008506.2 | 898 | 140 | ||
XM_015153021.2 | XP_015008507.1 | 555 | 140 | |||
XM_015153024.2 | XP_015008510.1 | 439 | 140 | |||
XM_028830026.1 | XP_028685859.1 | 832 | 140 | |||
XM_028830027.1 | XP_028685860.1 | 505 | 140 | |||
GenBank mRNA | JU335499.1 | - | 685 | 140 | ||
JU476440.1 | - | 548 | 140 | |||
JV656297.1 | - | 527 | 140 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
