Assay Details
Gene Symbol: TRUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266998.2 | NP_001253927.1 | 603 | 129 | ||
GenBank mRNA | JU333034.1 | - | 633 | 129 | ||
JU474380.1 | - | 603 | 129 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
