Assay Details
Gene Symbol: PRKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137330.2 | XP_014992816.2 | 1062 | 96 | ||
XM_015137331.2 | XP_014992817.2 | 1059 | 96 | |||
XM_015137332.2 | XP_014992818.2 | 1062 | 96 | |||
XM_028847918.1 | XP_028703751.1 | 1313 | 96 |
