Assay Details
Gene Symbol: NPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840046.1 | XP_028695879.1 | 2986 | 78 | ||
XM_028840047.1 | XP_028695880.1 | 2968 | 78 | |||
XM_028840048.1 | XP_028695881.1 | 2818 | 78 | |||
XM_028840049.1 | XP_028695882.1 | 2498 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
