Assay Details
Gene Symbol: RAB11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261789.1 | NP_001248718.1 | 633 | 154 | ||
GenBank mRNA | JU336914.1 | - | 628 | 154 | ||
JU477650.1 | - | 551 | 154 | |||
JV048581.1 | - | 633 | 154 | |||
JV664470.1 | - | 636 | 154 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
