Assay Details
Gene Symbol: B4GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103384.4 | XP_001103384.3 | 966 | 91 | ||
XM_028828502.1 | XP_028684335.1 | 966 | 91 | |||
GenBank mRNA | JU328610.1 | - | 6 - 7 | 771 | 91 | |
JV045290.1 | - | 1543 | 91 | |||
JV525954.1 | - | 6 - 7 | 752 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
