Assay Details
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267787.1 | NP_001254716.1 | 90 | 91 | ||
GenBank mRNA | JU332203.1 | - | 109 | 91 | ||
JU471587.1 | - | 90 | 91 | |||
JV045438.1 | - | 105 | 91 | |||
JV619748.1 | - | 129 | 91 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
