Assay Details
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266424.1 | NP_001253353.1 | 1160 | 86 | ||
XM_015111808.2 | XP_014967294.1 | 1295 | 86 | |||
GenBank mRNA | JU337218.1 | - | 935 | 86 | ||
JU477931.1 | - | 1160 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
