Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100101.4 | XP_001100101.2 | 745 | 149 | ||
XM_015111987.2 | XP_014967473.1 | 658 | 149 | |||
XM_015111988.2 | XP_014967474.1 | 546 | 149 | |||
XM_015111989.2 | XP_014967475.1 | 442 | 149 | |||
XM_015111990.2 | XP_014967476.1 | 658 | 149 | |||
XM_015111991.2 | XP_014967477.1 | 658 | 149 | |||
XM_015111992.2 | XP_014967478.1 | 745 | 149 | |||
XM_015111993.2 | XP_014967479.1 | 402 | 149 | |||
XM_015111994.2 | XP_014967480.1 | 658 | 149 | |||
XM_028831820.1 | XP_028687653.1 | 583 | 149 | |||
XM_028831821.1 | XP_028687654.1 | 583 | 149 | |||
XM_028831823.1 | XP_028687656.1 | 768 | 149 | |||
XM_028831824.1 | XP_028687657.1 | 583 | 149 | |||
XM_028831825.1 | XP_028687658.1 | 745 | 149 | |||
XM_028831826.1 | XP_028687659.1 | 658 | 149 | |||
GenBank mRNA | JU337229.1 | - | 629 | 149 | ||
JU472792.1 | - | 627 | 149 | |||
JV048781.1 | - | 649 | 149 | |||
JV563756.1 | - | 593 | 149 | |||
JV666245.1 | - | 656 | 149 |

More Information
Set Membership: |
![]() ![]() |
