Assay Details
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099660.4 | XP_001099660.2 | 734 | 156 | ||
GenBank mRNA | JV508332.1 | - | 5 - 6 | 701 | 156 | |
JV583190.1 | - | 5 - 6 | 692 | 156 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
