Assay Details
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260911.1 | NP_001247840.1 | 889 | 70 | ||
XM_015111925.2 | XP_014967411.1 | 1141 | 70 | |||
GenBank mRNA | JV043790.1 | - | 889 | 70 | ||
JV600895.1 | - | 1213 | 70 | |||
JV638256.1 | - | 889 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
