Assay Details
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266456.2 | NP_001253385.1 | 189 | 152 | ||
GenBank mRNA | JU320848.1 | - | 244 | 152 | ||
JV045745.1 | - | 190 | 152 |
Gene Symbol: PAPOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV636599.1 | - | 2493 | 152 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266456.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
