Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265893.1 | NP_001252822.1 | 5 - 6 | 1094 | 74 | |
XM_015116917.2 | XP_014972403.1 | 1358 | 74 | |||
XM_015116918.2 | XP_014972404.1 | 1163 | 74 | |||
XM_028834257.1 | XP_028690090.1 | 1117 | 74 | |||
GenBank mRNA | JU472522.1 | - | 5 - 6 | 1094 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
