Assay Details
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260611.1 | NP_001247540.1 | 3 - 4 | 343 | 111 | |
GenBank mRNA | JU320327.1 | - | 3 - 4 | 377 | 111 | |
JU472484.1 | - | 2 - 3 | 311 | 111 | ||
JV044998.1 | - | 3 - 4 | 343 | 111 | ||
JV639432.1 | - | 3 - 4 | 389 | 111 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
