Assay Details
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258091.1 | NP_001245020.1 | 77 | 128 | ||
XM_015138492.2 | XP_014993978.1 | 204 | 128 | |||
XM_028843894.1 | XP_028699727.1 | 204 | 128 | |||
GenBank mRNA | JU473704.1 | - | 77 | 128 |

More Information
Set Membership: |
![]() ![]() |
