Assay Details
Gene Symbol: GLYAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257270.1 | NP_001244199.1 | 295 | 151 | ||
XM_015114098.2 | XP_014969584.2 | 359 | 151 | |||
GenBank mRNA | CK231271.1 | - | 139 | 151 | ||
CO583152.1 | - | 304 | 151 | |||
JU478673.1 | - | 146 | 151 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
Panther Classification:
