Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115035.2 | XP_014970521.1 | 246 | 144 | ||
GenBank mRNA | JU332480.1 | - | 1 - 2 | 78 | 144 | |
JU471017.1 | - | 1 - 2 | 84 | 144 | ||
JV045892.1 | - | 1 - 2 | 96 | 144 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015115035.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
