Assay Details
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113359.1 | NP_001106830.1 | 4 - 5 | 772 | 155 | |
XM_015115035.2 | XP_014970521.1 | 1085 | 155 | |||
GenBank mRNA | EU124697.1 | - | 4 - 5 | 772 | 155 | |
JU332480.1 | - | 5 - 6 | 917 | 155 | ||
JU471017.1 | - | 5 - 6 | 923 | 155 | ||
JV045892.1 | - | 5 - 6 | 935 | 155 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
