Assay Details
Gene Symbol: PAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095275.4 | XP_001095275.3 | 285 | 127 | ||
XM_015149591.2 | XP_015005077.1 | 285 | 127 | |||
GenBank mRNA | JU487536.1 | - | 1 - 2 | 137 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() |
